Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak2 Mouse anti-Human, Clone: 691R5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266622

Catalog No. NBP266622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Jak2Monoclonalantibody specifically detects Jak2 in Human samples.
Specifications

Specifications

Jak2
691R5
Western Blot
Affinity Purified
JAK2
Recombinant full length human JAK2 protein expressed in E. coli.
Affinity purified
Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary
3717
Western Blot
Unconjugated
PBS (pH 7.2) with 1% BSA with 0.08% Sodium Azide
EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
Mouse
IgG2b
100μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit