Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak2 (p Tyr1007, p Tyr1008) Rabbit anti-Human, Clone: SY24-03, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP267429

Catalog No. NBP267429


Add to Cart

Description

Description

Jak2 Monoclonal specifically detects Jak2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Jak2 (p Tyr1007, p Tyr1008)
Monoclonal
1 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200
EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
Rabbit
Protein A purified
Cell Cycle and Replication, Phospho Specific, Signal Transduction
3717
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
SY24-03
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
JAK2
Synthetic phospho-peptide corresponding to residues surrounding Tyr1007 and 1008 of human JAK2.
100 μL
Primary
Human, Mouse, Rat
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit