Learn More
Aviva Systems Biology Corp JAK3 Antibody - middle region (ARP54306_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JAK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human JAK3. Alternate names: JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAKApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61168