Learn More
JAK3 Mouse anti-Human, Clone: OTI1B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812815

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Janus Activating Kinase (JAK) is a number of related tyrosine kinases involved in cytoplasmic signal transduction. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.Specifications
JAK3 | |
Monoclonal | |
Unconjugated | |
JAK3 | |
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK | |
Mouse | |
Affinity Chromatography | |
RUO | |
3718 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P52333 | |
JAK3 | |
Human recombit protein fragment corresponding to amino acids 811-1124 of human JAK3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.