Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JAKMIP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571124
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 93%.
JAKMIP3 gene ontology annotations related to this gene include kinase binding; microtubule binding.
Specifications
JAKMIP3 | |
Polyclonal | |
Unconjugated | |
JAKMIP3 | |
6330417G02Rik; AI593226; bA140A10.5; C10orf14; C10orf39; JAKMIP3; JAMIP3; Janus kinase and microtubule interacting protein 3; Janus kinase and microtubule interacting protein 3; janus kinase and microtubule-interacting protein 3; janus kinase and microtubule-interacting protein 3; Kiaa4091; mKIAA4091; NECC2; neuroendocrine long coiled-coil 2; neuroendocrine long coiled-coil protein 2; RGD1307177 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
282973 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VZ66 | |
JAKMIP3 | |
synthetic peptide directed towards the N-terminal region of human JAKMIP3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction