Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-A Rat anti-Mouse, DyLight 550, Clone: RM0116-1M21, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122494R
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot.Specifications
JAM-A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
F11R | |
Mouse recombinant protein of the extracellular domain of JAM-A | |
0.1 mL | |
50848 | |
Mouse | |
Purified |
Western Blot | |
RM0116-1M21 | |
Western Blot | |
CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
Rat | |
Protein A or G purified | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of extracellular domain of JAM-A. | |
Store at 4C in the dark. | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title