Learn More
JAM-B/VE-JAM Mouse anti-Human, Alexa Fluor™ 750, Clone: 988905, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10744S100UG
Catalog No. FB10744S100
Add to cart
This item is not returnable. View return policy
Description
Description
JAM-B/VE-JAM Monoclonal antibody specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
JAM-B/VE-JAM
988905
Flow Cytometry 0.25-1 μg/106 cells
P57087
Mouse
IgG2a
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human JAM/VE-JAM in direct ELISAs
Flow Cytometry
Alexa Fluor™ 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322
Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM, Phe29-Asn236, Accession No. P57087
100 μg
Primary
58494
Human