Learn More
Aviva Systems Biology Corp JAM3 Antibody - N-terminal region (ARP50121_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50121_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JAM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human JAM3. Alternate names: FLJ14529, JAM-C, JAMCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60262