Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAML Armenian Hamster anti-Mouse, Clone: eBio4E10 (4E10), eBioscience™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen 14566681

 View more versions of this product

Catalog No. 50-131-16


Add to cart

Description

Description

Description: The eBio4E10 monoclonal antibody reacts with mouse JAML (junctional adhesion molecule-like protein), also known as AMICA. JAML is a glycosylated membrane protein possessing two Ig-like domains in its extracellular domain. JAML is expressed on polymorphonuclear leukocytes (PMN), and is involved in the transmigration of such cells through epithelial tight junctions via interactions between JAML and epithelial coxsackie and adenovirus receptor (CAR). JAML is also expressed on lymphocyte subsets, such as gammadelta T cell lines, and can provide co-stimulatory signals for cell activation. Applications Reported: This eBio4E10 antibody has been reported for use in flow cytometric analysis and immunoprecipitation. Applications Tested: This eBio4E10 (4E10) antibody has been tested by flow cytometric analysis of mouse lysed whole blood. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

JAML (junctional adhesion molecule-like protein), also known as AMICA. JAML is a glycosylated membrane protein possessing two Ig-like domains in its extracellular domain. JAML is expressed on polymorphonuclear leukocytes (PMN), and is involved in the transmigration of such cells through epithelial tight junctions via interactions between JAML and epithelial coxsackie and adenovirus receptor (CAR). JAML is also expressed on lymphocyte subsets, such as gamma-delta T cell lines, and can provide co-stimulatory signals for cell activation.
Specifications

Specifications

JAML
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q80UL9
AMICA1, CREA7-1, JAML, adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Immunoprecipitation
eBio4E10 (4E10)
Unconjugated
Jaml
Liquid
Jaml
IgG
50 μg
4° C
Primary
270152
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.