Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Janice VanCleave Experimental Books
SCI_ED

Simply fascinating experiments.

$28.36 - $30.80

Specifications

Author Janice VanCleave
Grade 3 to 7
Format Book
Subject Science
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S79281
SCI_ED
Encompass
View Documents
JOHN WILEY & SONS
0471252239
203 Gooey, Slippery, Slimy, Weird and Fun Experiments
Each for $30.80
Add to cart
 
S79240
SCI_ED
Encompass
View Documents
JOHN WILEY & SONS
0471331015
204 Sticky, Gloppy, Wacky and Wonderful Experiments
Each for $28.36
Add to cart
 
Description

Description

Safe, simple experiments in astronomy, biology, chemistry, earth science, and physics. Many science principles are explained in simple and easy-to-understand terms and illustrations. Soft cover.

Order Info

Grades 3-7

Specifications

Specifications

Janice VanCleave
3 to 7
Book
Science
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit