Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jarid1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Jarid1A
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092658
View Documents
Novus Biologicals
NBP303856100UL
100 μg Each for $320.71
Add to cart
 
NB092659
View Documents
Novus Biologicals
NBP30385620UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Jarid1A Polyclonal antibody specifically detects Jarid1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

Jarid1A
Unconjugated
Rabbit
Human, Mouse, Rat
EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A
Recombinant protein of human Jarid1A.
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
5927
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit