Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JARID1C, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A301035AM

 View more versions of this product

Catalog No. 50-155-9073


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Dog and Pig

This gene is a member of the SMCY homolog family and encodes a protein with one ARID domain, one JmjC domain, one JmjN domain and two PHD-type zinc fingers. The DNA-binding motifs suggest this protein is involved in the regulation of transcription and chromatin remodeling. Mutations in this gene have been associated with X-linked mental retardation. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Specifications

Specifications

JARID1C
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
P41229
DXS1272E, JARID1C, MRXJ, MRXSCJ, MRXSJ, SMCX, XE169, JmjC domain-containing protein SMCX, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, Smcx homolog, X chromosome, Smcy homolog, X-linked, histone demethylase JARID1C, lysine-specific demethylase 5C, protein SmcX, sele
Rabbit
IgG
100 μl
4° C
Primary
8242
Western Blot
Unconjugated
KDM5C
liquid
KDM5C
Between 1510 and C-term
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit