Learn More
Thermo Scientific™ Javelin™ Direct-Connection Column Filters
Protect HPLC systems with one-piece Thermo Scientific™ Javelin Direct-Connection Column Filters.
$256.50 - $311.00
Specifications
For Use With (Equipment) HPLC Systems
Type Javelin Column Filter
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity & Availability  
88700
View Documents
Thermo Scientific™
88700
3 mm
Pack of 4 for $256.50
Add to Cart
88400
View Documents
Thermo Scientific™
88400
4.6/4.0 mm
Pack of 4 for $256.50
Add to Cart
88200
View Documents
Thermo Scientific™
88200
2.1 mm
Pack of 4 for $311.00
Add to Cart
Description
Description
Protect HPLC systems with one-piece Thermo Scientific™ Javelin Direct-Connection Column Filters.
  • Direct-connection design for maximum efficiency
  • Replace entire disposable filter unit for easy changes
  • Recommended for use as dedicated filters for a column rather than the HPLC system
  • 0.0625in. CPI tip attaches directly to HPLC column inlet without tubing or wrenches
  • 0.5μm porosity
Specifications
Specifications
HPLC Systems
Javelin Column Filter
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.