Learn More
JAZF1 293T Cell Overexpression Lysate (Denatured), Abnova
Supplier:  Abnova Corporation H00221895T01
Catalog No. 89-009-802
Add to Cart
Description
Description
Western Blotting
Specifications
Specifications
293T
tcag7.981 293T Cell Transient Overexpression Lysate(Denatured)
26.84
pCMV-tcag7.981 full-length
100 μL
Human
Western Blot
BC042441.1
6.8
Transient overexpression cell Lysate was tested with Anti-tcag7.981 antibody (H00221895-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations