Learn More
Invitrogen™ JDP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120335
Description
Positive test controls include: HeLa, Jurkat, HepG2, Rat testis. The target is usually found in the following locations: Nucleus. Immunogen sequence: MVAGWPATPP AMMPGQIPDP SVTTGSLPGL GPLTGLPSSA LTVEELKYAD IRNLGAMIAP LHFLEVKLGK RPQPVKSELD EEEERRKRRR EKNKVAAARC RNKKKERTEF LQRESERLEL MNAELKTQIE ELKQERQQLI LMLNRHRPTC IVRTDSVKTP ESEGNPLLEQ LEKK.
Component of the AP-1 transcription factor that represses transactivation mediated by the Jun family of proteins. Involved in a variety of transcriptional responses associated with AP-1 such as UV-induced apoptosis, cell differentiation, tumorigenesis and antitumogeneris. Can also function as a repressor by recruiting histone deacetylase 3/HDAC3 to the promoter region of JUN. May control transcription via direct regulation of the modification of histones and the assembly of chromatin.
Specifications
JDP2 | |
Polyclonal | |
Unconjugated | |
JDP2 | |
c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116674, 122953 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.54 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q78E65, Q8WYK2 | |
JDP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-174 of human JDP2 (NP_001128521.1). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.