Learn More
Smiths Medical Jelco™ Multi-Sample Blood Collection Needles
Smiths Medical Jelco™ Multi-Sample Blood Collection Needles with color-coded caps for quick and easy identification
Manufacturer:  Smiths Medical 110201020002
 View more versions of this product
Catalog No. 22-024-994
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Compatible with Venipuncture Needle-Pro™ devices
  • Available in a variety of gauges, and lengths
  • Color-coded caps and hubs allows rapid identification of needle gauge size
Specifications
Specifications
Black
Blood Collection
Venipuncture Needle-Pro™ Safety Holders
Sterile
No
22 ga
1 in.
Safety and Handling
Safety and Handling
ShelfLife : 5 Years