Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smiths Medical Jelco™ Multi-Sample Blood Collection Needles

Smiths Medical Jelco™ Multi-Sample Blood Collection Needles with color-coded caps for quick and easy identification

Specifications

Disposable No
For Use With (Application) Blood Collection
For Use With (Equipment) Venipuncture Needle-Pro™ Safety Holders
Sterility Sterile
View More Specs

Products 6
Catalog Number Mfr. No. Gauge Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Gauge Length (English) Price Quantity & Availability  
22-024-998
View Documents
Smiths Medical
110201020006
20 ga 1 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-024-999
View Documents
Smiths Medical
110201020007
20 ga 1.5 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-024-996
View Documents
Smiths Medical
110201020004
21 ga 1 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-024-997
View Documents
Smiths Medical
110201020005
21 ga 1.5 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-024-994
View Documents
Smiths Medical
110201020002
22 ga 1 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-024-995
View Documents
Smiths Medical
110201020003
22 ga 1.5 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
Description

Description

  • Compatible with Venipuncture Needle-Pro™ devices
  • Available in a variety of gauges, and lengths
  • Color-coded caps and hubs allows rapid identification of needle gauge size
Specifications

Specifications

No
Blood Collection
Venipuncture Needle-Pro™ Safety Holders
Sterile
Safety and Handling

Safety and Handling

ShelfLife : 5 Years

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit