Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OI Inc. Jencons™ Vac-Seal™ Glass Dewar Flasks

For storing liquid gases safely and easily

Specifications

For Use With (Application) Storage of liquid gases
Includes Lid, clamps
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Height (English) Height (Metric) Material Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Height (English) Height (Metric) Material Price Quantity & Availability  
03-692-122
View Documents
OI Inc.
238131
1 L 9.6 in. 24.38cm Metal Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. N/A
03-692-125
View Documents
OI Inc.
238147/DEL
1 L 9.6 in. 24.38cm Stainless Steel Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. N/A
03-692-126
View Documents
OI Inc.
238150/DEL
2 L 10.6 in. 26.92cm Stainless Steel Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. N/A
Description

Description

  • Vented, clamp-on lid with clips
  • Suitable for room-to-room transportation; unsuitable for vehicular transportation

  • 03-692-122 has a blue metal casing
  • 03-692-125 and 03-692-126 have a stainless-steel casing

Specifications

Specifications

Storage of liquid gases
Lid, clamps
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit