Learn More
Alfa Aesar™ Jenner's Stain
For use in staining blood smears
Manufacturer:  Alfa Aesar™ J6107722
 View more versions of this product
Catalog No. AAJ6107722
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
100g
For use in staining blood smears
62851-42-7
Hematology Stain
Powder
Dark Green
Eosin-Methylene Blue