Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jervine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
469-59-0
C27H39NO3
425.613
CLEXYFLHGFJONT-DNMILWOZSA-N
jervine,iervin,jerwiny polish,11-ketocyclopamine,unii-19v3ecx465,veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3beta,23beta,17,23beta-epoxy-3beta-hydroxyveratraman-11-one,spiro 9h-benzo a fluorene-9,2' 3'h-furo 3,2-b-pyridin-11 1h-one, 2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-hexadecahydro-3-hydroxy-3',6',10, 11b-tetramethyl,jerwiny,veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3-beta,23-beta
10098
CHEBI:6088
(3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3-hydroxy-3',6',10,11b-tetramethylspiro[1,2,3,4,6,6a,6b,7,8,11a-decahydrobenzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-11-one
CC1CC2C(C(C3(O2)CCC4C5CC=C6CC(CCC6(C5C(=O)C4=C3C)C)O)C)NC1
This item is not returnable. View return policy
469-59-0
C27H39NO3
425.613
CLEXYFLHGFJONT-DNMILWOZSA-N
jervine,iervin,jerwiny polish,11-ketocyclopamine,unii-19v3ecx465,veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3beta,23beta,17,23beta-epoxy-3beta-hydroxyveratraman-11-one,spiro 9h-benzo a fluorene-9,2' 3'h-furo 3,2-b-pyridin-11 1h-one, 2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-hexadecahydro-3-hydroxy-3',6',10, 11b-tetramethyl,jerwiny,veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3-beta,23-beta
10098
CHEBI:6088
(3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3-hydroxy-3',6',10,11b-tetramethylspiro[1,2,3,4,6,6a,6b,7,8,11a-decahydrobenzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-11-one
CC1CC2C(C(C3(O2)CCC4C5CC=C6CC(CCC6(C5C(=O)C4=C3C)C)O)C)NC1

Inhibitor of Hedgehog (Hh) signaling; binds directly to Smo

Inhibitor of Hedgehog signaling (IC50 = 500 - 700 nM); binds directly to Smoothened (Smo). Inhibits the response of target tissues to Sonic hedgehog signaling, possibly via the sterol sensing domain of Patched. Teratogenic.
Quantity 10mg
Formula Weight 425.6
Percent Purity >98%
Chemical Name or Material Jervine

Recommended Storage : Store at -20°C