Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Perfect for cell growth.
Description
- Flat, free flask surface
- Striation free to maximize usable growth area
- Used in both open, closed systems
- Angled neck design offers good pipet and cell scraper access
- Upper triangular, wider base shape provides stability
- Protruding ridge on back side of flask makes it easy for stacking
- Special area near neck for easy mark writing
- Both flask sides have engraved graduation
- Sterilized by gamma irradiation
- Non-pyrogenic
Warranty and Services
Three years
Specifications
CE | |
Natural | |
Cell growth | |
200 Flasks | |
Tissue Culture Flask | |
Flat | |
Sterile | |
Plug Seal |
Blue | |
50 mL | |
Angled | |
10 Flasks | |
Surface-treated | |
Yes | |
Flat | |
25 cm2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction