Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Jeweler Forceps, Sklar 4-1/2 in., Sharp, Straight, Smooth, #3, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 667447
Forceps, Jeweler Forceps, Sklar, 4-1/2 in., Sharp, Straight, Smooth, #3, EA. Sklar Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/2 inches. It is also known as the #3 style jeweler forceps. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-9030