Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP2 Mouse anti-Human, Mouse, Rat, Clone: S135-37, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527632

Catalog No. PIMA527632


Add to cart

Description

Description

The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.
Specifications

Specifications

JIP2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q13387, Q9ERE9
MAPK8IP2, C jun amino terminal kinase interacting protein 2, C-jun-amino-terminal kinase-interacting protein 2, Homologous to mouse JIP 1, IB 2, IB-2, IB2, Islet brain 2, Islet-brain-2, JIP 2, JIP-2, JIP2, JNK interacting protein 2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, MAPK8IP2, Mitogen activated protein kinase 8 interacting protein 2, Mitogen-activated protein kinase 8-interacting protein 2, PRKM8 interacting protein like, PRKM8IPL
Mouse
IgG1
100 μg
-20°C
Primary
23542, 315220, 60597
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S135-37
Unconjugated
MAPK8IP2
Liquid
MAPK8IP2
Fusion protein amino acids 226-421 of mouse JIP-2/IB-2. Rat: 94% identity (127/135 amino acids identical). Human: 86% identity (173/200 amino acids identical) >50% identity with JIP-1.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit