Learn More
JIP2 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S135-37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242194IR
Catalog No. NBP242194IR
Add to cart
Description
Description
JIP2 Monoclonal antibody specifically detects JIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
JIP2
S135-37
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
MAPK8IP2
Fusion protein amino acids 226-421 of mouse JIP-2/IB-2. Rat: 94% identity (127/135 amino acids identical). Human: 86% identity (173/200 amino acids identical) >50% identity with JIP-1.
Protein G purified
Apoptosis
Primary
23542
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
C-Jun-amino-terminal kinase-interacting protein 2, homologous to mouse JIP-1, IB-2, IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2, islet-brain 2, Islet-brain-2, JIP-2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, mitogen-activated protein kinase 8 interacting protein 2, PRKM8 interacting protein-like, PRKM8IPL
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then a 50% identity with JIP-1.Detects approx Greater then 100 kDa. Does not cross-react with JIP-1/IB-1.