Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP2 Mouse anti-Human, Mouse, Rat, FITC, Clone: S135-37, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242194F

Catalog No. NBP242194F


Add to cart

Description

Description

JIP2 Monoclonal antibody specifically detects JIP2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

JIP2
S135-37
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
MAPK8IP2
Fusion protein amino acids 226-421 of mouse JIP-2/IB-2. Rat: 94% identity (127/135 amino acids identical). Human: 86% identity (173/200 amino acids identical) >50% identity with JIP-1.
Protein G purified
Apoptosis
Primary
23542
Human, Mouse, Rat
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
C-Jun-amino-terminal kinase-interacting protein 2, homologous to mouse JIP-1, IB-2, IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2, islet-brain 2, Islet-brain-2, JIP-2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, mitogen-activated protein kinase 8 interacting protein 2, PRKM8 interacting protein-like, PRKM8IPL
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then a 50% identity with JIP-1.Detects approx Greater then 100 kDa. Does not cross-react with JIP-1/IB-1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit