Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31000025UL

 View more versions of this product

Catalog No. NB124902


Add to cart

Description

Description

JIP2 Polyclonal antibody specifically detects JIP2 in Human samples. It is validated for Western Blot
Specifications

Specifications

JIP2
Unconjugated
PBS buffer, 2% sucrose
C-Jun-amino-terminal kinase-interacting protein 2, homologous to mouse JIP-1, IB-2, IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2, islet-brain 2, Islet-brain-2, JIP-2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, mitogen-activated protein kinase 8 interacting protein 2, PRKM8 interacting protein-like, PRKM8IPL
The immunogen is a synthetic peptide directed towards the middle region of human JIP2 (NP_036456.1). Peptide sequence GTSPSSDPGIEADLRSRSSGGRGGRRSSQELSSPGSDSEDAGGARLGRMI
25 μg
Apoptosis
Primary
23542
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit