Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JLK 6, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
62252-26-0
C10H8ClNO3
225.628
AMDGKLWVCUXONP-UHFFFAOYSA-N
7-amino-4-chloro-3-methoxyisocoumarin, jlk 6, jlk6, 7-amino-4-chloro-3-methoxy-isocoumarin, gamma-secretase inhibitor xi, 1h-2-benzopyran-1-one, 7-amino-4-chloro-3-methoxy, jlk-6, 3-methoxy-4-chloro-7-aminoisocoumarin, 7-amino-3-methoxy-4-chloroisocoumarin, 7-amino-4-chloro-3-methoxy-1h-2-benzopyran
3803
7-amino-4-chloro-3-methoxyisochromen-1-one
COC1=C(C2=C(C=C(C=C2)N)C(=O)O1)Cl
This item is not returnable. View return policy
62252-26-0
C10H8ClNO3
225.628
AMDGKLWVCUXONP-UHFFFAOYSA-N
7-amino-4-chloro-3-methoxyisocoumarin, jlk 6, jlk6, 7-amino-4-chloro-3-methoxy-isocoumarin, gamma-secretase inhibitor xi, 1h-2-benzopyran-1-one, 7-amino-4-chloro-3-methoxy, jlk-6, 3-methoxy-4-chloro-7-aminoisocoumarin, 7-amino-3-methoxy-4-chloroisocoumarin, 7-amino-4-chloro-3-methoxy-1h-2-benzopyran
3803
7-amino-4-chloro-3-methoxyisochromen-1-one
COC1=C(C2=C(C=C(C=C2)N)C(=O)O1)Cl

Inhibitor of γ-secretase-mediated βAPP processing

Inhibitor of γ-secretase that selectively inhibits βAPP cleavage without affecting other γ-secretase-mediated pathways. Prevents recovery of Aβ40 and Aβ42 from HEK293 cell overexpressing wild-type or Swedish-mutated βAPP (IC50 ∽ 30 μM) but displays no effect on Notch cleavage and Notch-mediated intracellular signaling. Displays no activity on BACE1, BACE2, α-secretase, the proteosome or GSK3β.
Quantity 50 mg
Formula Weight 225.63
Percent Purity >99%
Chemical Name or Material JLK 6

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit