Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2B / KDM4B Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody

Manufacturer:  Active MotifSupplier Diversity Partner 61221

Encompass

 View more versions of this product

Catalog No. 50-199-3246


Add to cart

Description

Description

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
Specifications

Specifications

JMJD2B / KDM4B
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
KDM4B
Liquid
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
23030
KDM4B
Lot-Specific
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
O94953
4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; Jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; Lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B
This JMJD2B / kDM4B antibody was raised against a peptide within the C-terminal region of human JMJD2B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit