Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2B Mouse anti-Human, Mouse, Clone: 3A6H6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261800
Description
JMJD2B Monoclonal antibody specifically detects JMJD2B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
JMJD2B | |
3A6H6 | |
Unconjugated | |
Purified | |
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B | |
KDM4B | |
Purified recombinant fragment of human KDM4B (AA: 960-1096) expressed in E. Coli. | |
122 kDa | |
Primary | |
0.1 ml | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot,Flow Cytometry,ELISA | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
PBS with 0.05% Sodium Azide | |
23030 | |
Mouse | |
IgG1 | |
Monoclonal | |
Protein G purified | |
Chromatin Modifiers, Chromatin Research, Epigenetics | |
Human,Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title