Learn More
JMJD2B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259196
Catalog No. NBP259196
Add to cart
Description
Description
JMJD2B Polyclonal antibody specifically detects JMJD2B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence.
Specifications
Specifications
JMJD2B
Unconjugated
Serum with 0.05% Sodium Azide
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B
Rabbit
IgG
100 ul
Chromatin Modifiers, Chromatin Research, Epigenetics
Primary
23030
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Western Blot 1:1000, ELISA 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunofluorescence 1:200
Antiserum
KDM4B
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human