Learn More
Abnova™ JMJD2D 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055693T01
Catalog No. 89-949-435
Add to cart
Specifications
Specifications
293T
NM_018039
pCMV-JMJD2D full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
JMJD2D 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 58.6kDa
Transient overexpression cell lysate was tested with anti-JMJD2D antibody (Mfr. No. H00055693-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human