Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | JMJD5 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
JMJD5 Polyclonal specifically detects JMJD5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
JMJD5 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
79831 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 1.14.11.27, FLJ13798, JmjC domain-containing protein 5, jumonji domain containing 5, Jumonji domain-containing protein 5, KDM8, lysine-specific demethylase 8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 147-416 of human JMJD5 (NP_079049.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title