Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD8 Antibody, Janelia Fluor™ 585, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177076JF585
Description
JMJD8 Polyclonal antibody specifically detects JMJD8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
JMJD8 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
339123 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
Janelia Fluor 585 | |
jmjC domain-containing protein 8, C16orf20, jumonji domain containing 8, PP14397 | |
Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human JMJD8. The immunogen is located within amino acids 250 - 300 of JMJD8. Amino Acid Squence: FLYPPEKTPEFHPNKTTL | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction