Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNJ 1661010, Tocris Bioscience™

Catalog Number p-4784056
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
681136-29-8
C19H19N5OS
365.46
MFCD00209157
BHBOSTKQCZEAJM-UHFFFAOYSA-N
n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide, 1-piperazinecarboxamide, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl-1-piperazinecarboxamide, acmc-20emff, d0w5wg, jnj hplc, n1-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide
2809273
N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
O=C(NC1=CC=CC=C1)N1CCN(CC1)C1=NC(=NS1)C1=CC=CC=C1
This item is not returnable. View return policy
681136-29-8
C19H19N5OS
365.46
MFCD00209157
BHBOSTKQCZEAJM-UHFFFAOYSA-N
n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide, 1-piperazinecarboxamide, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl-1-piperazinecarboxamide, acmc-20emff, d0w5wg, jnj hplc, n1-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide
2809273
N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
O=C(NC1=CC=CC=C1)N1CCN(CC1)C1=NC(=NS1)C1=CC=CC=C1

Selective, reversible FAAH inhibitor

Selective, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 = 12 nM). Brain penetrant and active in vivo.
Quantity 10 mg
Formula Weight 365.45
Percent Purity >99%
Chemical Name or Material JNJ 1661010
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit