Tocris™ JNJ 27141491

Potent and selective human CCR2 antagonist

Manufacturer: Tocris™ 5176/10

 View all options for this product

Catalog No. 51-761-0

  Check Availability
Add to Cart


For Research Use Only


    Description JNJ 27141491
    Molecular Weight 379.38
    Purity >98
    Quantity 10mg
    Product Type JNJ 27141491

    Potent, noncompetitive human CCR2 antagonist; inhibits MCP-1-induced Ca2+ mobilization in hCCR2-CHO cells (IC50 = 13nM). Exhibits selectivity for CCR2 over CCR1, CCR3-8 and CXCR1-3. Delays onset and reduces neurological signs in an experimental autoimmune encephalomyelitis model of multiple sclerosis in hCCR2 knock-in mice. Orally active.

    • Chemical Name: 3-[(1S)-1-(3,4-Difluorophenyl)propyl]-2,3-dihydro-5-(5-isoxazolyl)-2-thioxo-1H-imidazole-4-carboxylic acid methyl ester
    • Formula: C17H15F2N3O3S
    • CAS Number: 871313-59-6
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit