Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human JNK1 His Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP2521230.1MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-427 of Human JNK1 The Recombinant Human JNK1 Protein is derived from E. coli. The Recombinant Human JNK1 Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate Buffer Saline (pH7.4) containing 20% glycerol, 1mM DTT | |
TMW: 50.5kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
JNK1 | |
Recombinant |
SDS-PAGE | |
5599 | |
0.1mg | |
c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
Unconjugated | |
>85%, by SDS-PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction