Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK2 Rabbit anti-Human, Clone: 4, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | JNK2 |
---|---|
Clone | 4 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 0.2-1 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
JNK2 Monoclonal specifically detects JNK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
JNK2 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 0.2-1 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human | |
c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2 | |
Recombinant Human JNK2 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
4 | |
Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
0.2 um filtered solution in PBS | |
5601 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title