Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
JNK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 173371AP
Catalog No. 50-173-1517
Description
Description
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.
Specifications
Specifications
JNK2
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P45984, P49186, Q9WTU6
c Jun N terminal kinase 2, JNK 55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, MAP kinase 9, MAPK 9, MAPK9, p54a, p54aSAPK, PRKM9, SAPK
Rabbit
IgG
150 μL
-20°C
Primary
26420, 50658, 5601
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MAPK9
Liquid
Mapk9
JNK2 Fusion Protein Ag11372
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat