Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Josephin1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271506AF700

Catalog No. N271506A700


Add to cart

Description

Description

Josephin1 Monoclonal antibody specifically detects Josephin1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Josephin1
OTI3B11
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
JOSD1
Full length recombinant protein of human Josephin1 (NP_055691) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.4.19.12, Josephin domain containing 1, Josephin domain-containing 1, Josephin-1, JSPH1, KIAA0063dJ508I15.2
Mouse
IgG1
0.1 mL
Primary
9929.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit