Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JPH2 Monoclonal Antibody (OTI2D3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526300
Description
JPH2 Monoclonal antibody specifically detects JPH2 in Human samples. It is validated for Western Blot.Specifications
JPH2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BR39 | |
JPH2 | |
Full length human recombinant protein of JPH2 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI2D3 | |
Unconjugated | |
JPH2 | |
1110002E14Rik; CMH17; Jp2; JP-2; JP2NT; JPH2; jph2 {ECO:0000312; junctophilin 2; junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment; RGD:1305196} | |
Mouse | |
Affinity Chromatography | |
RUO | |
57158 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction