Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JRKL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548934

Catalog No. PA548934

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of this gene has not yet been defined, however, the encoded protein shares similarity with the human (41% identical) and mouse (34% identical) jerky gene products. This protein may act as a nuclear regulatory protein. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, May 2012]
Specifications

Specifications

JRKL
Polyclonal
PBS with 0.09% sodium azide
Q9Y4A0
Jerky protein homolog-like, Human homolog of mouse jerky gene protein, HHMJG, JRKL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8690
Western Blot
Unconjugated
JRKL
Liquid
JRKL
KLH conjugated synthetic peptide between 412-439 amino acids from the C-terminal region of human JRKL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit