Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Julolidine, 98%

Click to view available options
Quantity:
1g
5g
25g
479-59-4
C12H15N
173.259
MFCD00006917
DZFWNZJKBJOGFQ-UHFFFAOYSA-N
julolidine,2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine,1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro,1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline,2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine,2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline,2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
68069
C1CC2=C3C(=CC=C2)CCCN3C1
479-59-4
C12H15N
173.259
MFCD00006917
DZFWNZJKBJOGFQ-UHFFFAOYSA-N
julolidine,2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine,1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro,1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline,2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine,2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline,2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
68069
C1CC2=C3C(=CC=C2)CCCN3C1
Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
Flash Point >110°C (230°F)
Quantity 5g
Boiling Point 170°C to 173°C (31mmHg)
Melting Point 32°C to 37°C
Beilstein 139925
Formula Weight 173.26
Percent Purity 98%
Chemical Name or Material Julolidine

TSCA : TSCA

Recommended Storage : 2° to 8°C