Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Julolidine hydrobromide, 97%, Thermo Scientific™

Catalog Number AAH3629814
Click to view available options
Quantity:
5 g
25 g
100 g
83646-41-7
C12H16BrN
254.17
MFCD00198009
KHWBRFVBPGPEOJ-UHFFFAOYSA-N
julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
12255670
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
Br.C1CN2CCCC3=C2C(C1)=CC=C3
83646-41-7
C12H16BrN
254.17
MFCD00198009
KHWBRFVBPGPEOJ-UHFFFAOYSA-N
julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
12255670
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
Br.C1CN2CCCC3=C2C(C1)=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 239°C to 242°C
Color Pink
Quantity 25 g
UN Number UN2923
Sensitivity Hygroscopic
Formula Weight 254.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Julolidine hydrobromide
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit