Alfa Aesar™ Julolidine hydrobromide, 97%

Manufacturer: Alfa Aesar™ H3629822

Quantity 100g

 View more versions of this product

Catalog No. AAH3629822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 83646-41-7
    Formula Weight 254.17
    MDL Number MFCD00198009
    UN Number UN2923
    Synonym 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine hydrobromide
    Chemical Name or Material Julolidine hydrobromide

    Specifications

    Formula Weight 254.17
    Melting Point 239° to 242°C
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00198009
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Sensitivity Hygroscopic
    Physical Form Solid
    CAS 83646-41-7
    Synonym 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine hydrobromide
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Color Pale pink
    UN Number UN2923
    Quantity 100g
    Chemical Name or Material Julolidine hydrobromide
    Assay 97%

    GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit