Learn More
Andwin Scientific JUMBO DISPLAY ALARM TIMER/STOP
SDP
Supplier:  Andwin Scientific WT24706
Encompass_Preferred
Jumbo Display Alarm Timer/Stopwatch, Four independently programmable channels count up or down for any interval up to 100 hr., Each channel has a distinct 60-second audible alarm, Package: EA
Catalog No. NC9187574