Learn More
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
$787.00
Specifications
Material (Barrel) Acrylic
For Use With (Application) For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
SG009910
View Documents
Trajan™
009910
500 mL Each for $787.00
Add to cart
Description
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Specifications
Acrylic
For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling