Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jumonji/JARID2 Rabbit anti-Human, Mouse, Rat, Clone: 3L5C5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Jumonji/JARID2 |
---|---|
Clone | 3L5C5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Jumonji/JARID2 Monoclonal antibody specifically detects Jumonji/JARID2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Jumonji/JARID2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3720 | |
IgG | |
Affinity purified |
3L5C5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Jumonji/JARID2 (Q92833). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title