Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunB/AP-1 Mouse anti-Human, Clone: PCRP-JUNB-3G11, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen JunB/AP-1
Clone PCRP-JUNB-3G11
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122630
View Documents
Novus Biologicals
NBP30757820UG
20 μg
Each for $138.37
Add to cart
 
NB122629
View Documents
Novus Biologicals
NBP30757810UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

JunB/AP-1
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
Human
activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B
Recombinant full-length human JunB/AP-1 protein
Monoclonal
Protein A or G purified
PCRP-JUNB-3G11
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
3726
IgG2c
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit