Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUP Mouse anti-Chicken, Human, Clone: 15F11, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14827

Catalog No. 89-034-910


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant chicken JUP.

  • High specificity
  • High reactivity
Specifications

Specifications

JUP
15F11
Mouse monoclonal antibody raised against full length recombinant chicken JUP.
In 10 mM PBS.
Liquid
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Chicken, Human
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
E1C1V3
LOC429710
Recombinant protein corresponding to full length chicken JUP.
Protein A/G purification
RUO
Primary
429710
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)