Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUP Rabbit anti-Bovine, Cat, Chicken, Dog, Human, Mouse, Rat, Unconjugated, Clone: CTNG/2155R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00565

Catalog No. 89-339-521


Add to cart

Description

Description

JUP Monoclonal antibody specifically detects JUP in Bovine, Cat, Chicken, Dog, Human, Mouse, Rat samples. It is validated for WB, ELISA, IF, Flow Cyt.
Specifications

Specifications

JUP
CTNG/2155R
junction plakoglobin
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Bovine, Feline, Chicken, Dog, Human, Mouse, Rat
Western Blot, ELISA, Immunofluorescence, Flow Cytometry
Unconjugated
JUP
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
Recombinant protein corresponding to full-length human JUP
100 μg
Primary
3728
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit